Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003812.3 | NP_003803.1 | 12 | 1447 | ||
XM_005246932.3 | XP_005246989.1 | 12 | 1547 | |||
XM_011512086.2 | XP_011510388.1 | 12 | 1547 | |||
GenBank mRNA | AB009672.1 | - | 12 | 1447 | ||
AJ005580.1 | - | 12 | 1276 | |||
AK129906.1 | - | 8 | 611 | |||
AK296844.1 | - | 12 | 1156 | |||
BC132763.1 | - | 12 | 1427 | 40146785 | ||
BC132765.1 | - | 12 | 1427 | 40146787 |