Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMPCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004279.2 | NP_004270.2 | 10 | 1230 | ||
XM_005250717.3 | XP_005250774.1 | 10 | 1244 | |||
XM_006716181.3 | XP_006716244.1 | 10 | 1243 | |||
GenBank mRNA | AF054182.1 | - | 10 | 1209 | ||
AK000994.1 | - | 10 | 1212 | |||
AK057656.1 | - | 9 | 1028 | |||
AK090763.1 | - | 1 | 1107 | |||
AK297354.1 | - | 10 | 1595 | |||
AL050216.1 | - | 6 | 679 | |||
BC014079.2 | - | 10 | 1170 |