Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177608.1 | NP_001171079.1 | 2 | 377 | ||
NM_001772.3 | NP_001763.3 | 2 | 377 | |||
XM_011527531.2 | XP_011525833.1 | 8 | 1451 | |||
XM_011527532.2 | XP_011525834.1 | 7 | 1414 | |||
XM_017027508.1 | XP_016882997.1 | 8 | 1587 | |||
XM_017027509.1 | XP_016882998.1 | 7 | 1539 | |||
GenBank mRNA | AK304810.1 | - | 2 | 341 | ||
AK309558.1 | - | 2 | 358 | |||
BC028152.1 | - | 2 | 342 | 5217182 | ||
BP295158.1 | - | 2 | 377 | |||
BP303860.1 | - | 2 | 379 | |||
M23197.1 | - | 2 | 349 |