Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004995.3 | NP_004986.1 | 4 | 964 | ||
GenBank mRNA | AK291325.1 | - | 4 | 906 | ||
AK312657.1 | - | 4 | 912 | |||
BC064803.1 | - | 4 | 898 | 6046773 | ||
D26512.1 | - | 4 | 790 | |||
HE978355.1 | - | 4 | 913 | |||
U41078.1 | - | 4 | 679 | |||
X83535.1 | - | 4 | 798 | |||
X90925.1 | - | 4 | 791 | |||
Z48481.1 | - | 4 | 792 |