Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFG3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006796.2 | NP_006787.2 | 14, 15 | 1981 | ||
GenBank mRNA | AK129504.1 | - | 7, 8 | 807 | ||
BC024282.1 | - | 15, 16 | 1834 | |||
BC065016.1 | - | 14, 15 | 1903 | 6169804 | ||
Y18314.1 | - | 14, 15 | 1900 |