Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317183.1 | NP_001304112.1 | 3 | 467 | ||
NM_006838.3 | NP_006829.1 | 4 | 536 | |||
NR_133673.1 | 4 | 536 | ||||
XM_005268583.3 | XP_005268640.1 | 4 | 455 | |||
GenBank mRNA | AK055692.1 | - | 258 | |||
AK091730.1 | - | 3 | 343 | |||
AK125296.1 | - | 4 | 1062 | |||
AK315559.1 | - | 4 | 473 | |||
BC013782.1 | - | 4 | 414 | 4340717 | ||
U13261.1 | - | 4 | 436 |