Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006988.4 | NP_008919.3 | 7 | 2332 | ||
GenBank mRNA | AB037767.1 | - | 7 | 1992 | ||
AF060152.1 | - | 7 | 2063 | |||
AF170084.1 | - | 7 | 2119 | |||
AF207664.1 | - | 7 | 2319 | |||
AK023795.1 | - | 3 | 1426 | |||
AK314475.1 | - | 7 | 2332 | |||
AL162080.1 | - | 4 | 626 | |||
BC036515.2 | - | 7 | 2332 | 5266513 | ||
BX647388.1 | - | 7 | 2331 |