Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007038.4 | NP_008969.2 | 8 | 3490 | ||
XM_011529448.2 | XP_011527750.1 | 7 | 3322 | |||
GenBank mRNA | AF086299.1 | - | 1 | 35 | ||
AF141293.1 | - | 6 | 1525 | |||
AF142099.1 | - | 8 | 2883 | |||
BC093775.1 | - | 8 | 2845 | 7939620 | ||
BC093777.1 | - | 8 | 2845 | 7939622 |