Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320313.1 | NP_001307242.1 | 12 | 1269 | ||
NM_014237.2 | NP_055052.1 | 13 | 1341 | |||
NR_135201.1 | 11 | 979 | ||||
GenBank mRNA | AJ133004.1 | - | 13 | 1286 | ||
AK313961.1 | - | 13 | 1331 | |||
AY358321.1 | - | 12 | 1256 | |||
BC121045.1 | - | 13 | 1286 | 40117918 | ||
BC121046.1 | - | 11 | 924 |