Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130703.1 | NP_001124175.1 | 4 | 1739 | ||
NM_001130704.1 | NP_001124176.1 | 4 | 1714 | |||
NM_001130705.1 | NP_001124177.1 | 3 | 1633 | |||
NM_001278125.1 | NP_001265054.1 | 6 | 1881 | |||
NM_001278126.1 | NP_001265055.1 | 6 | 1874 | |||
NM_001278127.1 | NP_001265056.1 | 2 | 1444 | |||
NM_014269.4 | NP_055084.3 | 5 | 1820 | |||
XM_011531556.1 | XP_011529858.1 | 6 | 1963 | |||
XM_011531557.1 | XP_011529859.1 | 7 | 2121 | |||
XM_011531559.1 | XP_011529861.1 | 5 | 1852 | |||
XM_011531560.1 | XP_011529862.1 | 5 | 1879 | |||
XM_011531561.1 | XP_011529863.1 | 4 | 1771 | |||
GenBank mRNA | AF134708.1 | - | 2 | 1528 | ||
AF171929.1 | - | 1 | 1150 | |||
AF171930.1 | - | 1 | 1150 | |||
AK292410.1 | - | 4 | 1669 |