Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NETO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018092.4 | NP_060562.3 | 5 | 926 | ||
XM_006721289.1 | XP_006721352.1 | 5 | 926 | |||
XM_006721292.1 | XP_006721355.1 | 5 | 926 | |||
XM_017023740.1 | XP_016879229.1 | 5 | 640 | |||
GenBank mRNA | AK074937.1 | - | 5 | 898 | ||
AL834404.1 | - | 2 | 292 | |||
AY358718.1 | - | 5 | 823 | |||
BC065504.1 | - | 5 | 607 | |||
BX538063.1 | - | 3 | 412 |