Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPXM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184699.1 | NP_001171628.1 | 13 | 1756 | ||
NM_019609.4 | NP_062555.1 | 13 | 1978 | |||
GenBank mRNA | AK027661.1 | - | 13 | 1914 | ||
AK075508.1 | - | 9 | 1190 | |||
AK075527.1 | - | 13 | 1687 | |||
AK303142.1 | - | 10 | 1457 | |||
AK315027.1 | - | 13 | 1743 | |||
AY358956.1 | - | 13 | 1934 | |||
BC063430.1 | - | 13 | 1955 | 5239864 |