Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319182.1 | NP_001306111.1 | 7 | 997 | ||
NM_001319183.1 | NP_001306112.1 | 6 | 1283 | |||
NM_001319184.1 | NP_001306113.1 | 6 | 1137 | |||
NM_020216.3 | NP_064601.3 | 7 | 1286 | |||
GenBank mRNA | AJ242586.1 | - | 7 | 1287 | ||
AJ296161.1 | - | 7 | 1257 | |||
AK026644.1 | - | 4 | 489 | |||
AK097372.1 | - | 8 | 1200 | |||
AL390139.1 | - | 6 | 966 | |||
BC001064.2 | - | 7 | 1271 | 2823082 | ||
BC012166.1 | - | 7 | 1263 | 4636934 | ||
Z36876.1 | - | 2 | 111 |