Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001785.2 | NP_001776.1 | 2 | 419 | ||
GenBank mRNA | AJ000474.1 | - | 2 | 246 | ||
BC048284.1 | - | 3 | 542 | |||
BC054036.1 | - | 2 | 304 | 5403243 | ||
CD675458.1 | - | 2 | 407 | |||
L27943.1 | - | 2 | 357 | |||
S52873.1 | - | 2 | 237 |