Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT7 SIRNA Assays ›
Gene Symbol:
SEPT7
Gene Name:
septin 7
Gene Aliases:
CDC10, CDC3, NBLA02942, SEPT7A
Chromosome Location:
Chr.7: 35800986 - 35915763 on Build GRCh38
UniGene ID:
Hs.191346
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
10417
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001011553.3
NP_001011553.2
3
473
NM_001242956.1
NP_001229885.1
2
368
NM_001788.5
NP_001779.3
4
476
XM_006715806.3
XP_006715869.1
4
424
XM_011515656.2
XP_011513958.1
4
424
XM_011515661.2
XP_011513963.1
4
424
XM_017012855.1
XP_016868344.1
4
408
XM_017012856.1
XP_016868345.1
4
416
XM_017012857.1
XP_016868346.1
4
413
XM_017012858.1
XP_016868347.1
5
338
XM_017012859.1
XP_016868348.1
4
335
XM_017012860.1
XP_016868349.1
5
332
XM_017012861.1
XP_016868350.1
5
329
XM_017012862.1
XP_016868351.1
3
239
XM_017012863.1
XP_016868352.1
4
782
GenBank mRNA
AB073389.1
-
4
243
AB209677.1
-
4
211
AK223225.1
-
4
243
AK223241.1
-
4
243
AK290545.1
-
3
314
AK297883.1
-
2
135
AK300601.1
-
5
189
BC025987.2
-
3
240
4823810
BC067264.1
-
3
219
6503495
BC093640.1
-
4
242
7939485
BC093642.1
-
4
242
7939487
BC107718.1
-
3
421
BI669760.1
-
4
454
BP280069.1
-
4
447
BQ229116.1
-
3
404
BQ232357.1
-
4
218
BX571755.1
-
3
241
DC414394.1
-
3
476
DQ232879.1
-
3
224
KF305319.1
-
5
308
S72008.1
-
4
243
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cytokinesis
mitotic nuclear division
regulation of embryonic cell shape
protein heterooligomerization
cilium morphogenesis
cell-cell adhesion
positive regulation of nonmotile primary cilium assembly
structural molecule activity
protein binding
GTP binding
identical protein binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-zpscg:80/10.239.160.225:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0