Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001792.4 | NP_001783.2 | 2 | 573 | ||
XM_011525788.1 | XP_011524090.1 | 2 | 238 | |||
XM_017025514.1 | XP_016881003.1 | 2 | 544 | |||
GenBank mRNA | BC036470.1 | - | 2 | 265 | 5264944 | |
CN263272.1 | - | 2 | 281 | |||
DA225616.1 | - | 2 | 386 | |||
S42303.1 | - | 2 | 607 | |||
X54315.1 | - | 2 | 250 |