Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321065.1 | NP_001307994.1 | 4 | 611 | ||
NM_001321066.1 | NP_001307995.1 | 5 | 673 | |||
NM_001645.4 | NP_001636.1 | 4 | 547 | |||
GenBank mRNA | AK225971.1 | - | 4 | 533 | ||
BC009698.2 | - | 3 | 370 | 3889900 | ||
BC055093.1 | - | 5 | 599 | 6567705 | ||
X00570.1 | - | 3 | 403 |