Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313994.1 | NP_001300923.1 | 15 | 1796 | ||
NM_145004.6 | NP_659441.3 | 18 | 2194 | |||
XM_006716298.2 | XP_006716361.1 | 18 | 1988 | |||
XM_006716299.1 | XP_006716362.1 | 16 | 1670 | |||
XM_011544433.1 | XP_011542735.1 | 18 | 1988 | |||
XM_011544434.1 | XP_011542736.1 | 16 | 1769 | |||
XM_017013183.1 | XP_016868672.1 | 17 | 1919 | |||
XM_017013184.1 | XP_016868673.1 | 18 | 1988 | |||
XM_017013185.1 | XP_016868674.1 | 17 | 1994 | |||
XM_017013186.1 | XP_016868675.1 | 14 | 1598 | |||
XM_017013187.1 | XP_016868676.1 | 10 | 1134 | |||
XM_017013188.1 | XP_016868677.1 | 9 | 1103 | |||
GenBank mRNA | AK122608.1 | - | 16 | 1549 | ||
AK129810.1 | - | 4 | 1096 | |||
AK131038.1 | - | 10 | 1163 | |||
AK300148.1 | - | 15 | 1754 | |||
AK313864.1 | - | 18 | 2015 | |||
AY172952.1 | - | 16 | 1798 | |||
AY358739.1 | - | 18 | 2074 | |||
BC026085.2 | - | 18 | 1978 | 4838404 | ||
BC026169.2 | - | 18 | 2005 | 4829390 | ||
BC028702.2 | - | 18 | 1988 | 4829385 | ||
BC030014.2 | - | 18 | 1981 | 4830659 | ||
BC030698.2 | - | 18 | 1996 | 4822988 | ||
BC034975.2 | - | 18 | 1989 | 4829806 |