Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LVRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173800.4 | NP_776161.3 | 14, 15 | 2421 | ||
GenBank mRNA | AK075131.1 | - | 9, 10 | 956 | ||
AK291655.1 | - | 14, 15 | 2435 | |||
AY560010.1 | - | 14, 15 | 2309 | |||
BC036440.1 | - | 9, 10 | 1300 | |||
BC060869.1 | - | 15, 16 | 2348 | 30346049 | ||
BC070028.1 | - | 9, 10 | 1418 | |||
BC094716.1 | - | 9, 10 | 1418 | |||
BC109022.1 | - | 14, 15 | 2285 | 40008501 | ||
BC109023.1 | - | 14, 15 | 2285 | 40008502 |