Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001150.2 | NP_001141.2 | 21 | 3240 | ||
XM_005254892.4 | XP_005254949.1 | 21 | 3248 | |||
XM_011521473.1 | XP_011519775.1 | 21 | 3391 | |||
GenBank mRNA | AB209918.1 | - | 21 | 3236 | ||
AK298134.1 | - | 15 | 1710 | |||
AK298242.1 | - | 15 | 1761 | |||
AK298339.1 | - | 16 | 1821 | |||
AK300946.1 | - | 17 | 1971 | |||
BC058928.1 | - | 20 | 3041 | 6020902 | ||
M22324.1 | - | 20 | 3068 | |||
X13276.1 | - | 20 | 2972 |