Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272101.1 | NP_001259030.1 | 7 | 1938 | ||
NM_002429.5 | NP_002420.1 | 9 | 2206 | |||
NR_073606.1 | 9 | 2283 | ||||
XM_006719401.3 | XP_006719464.1 | 7 | 1847 | |||
XM_011538359.1 | XP_011536661.1 | 8 | 1946 | |||
GenBank mRNA | AK225939.1 | - | 6 | 1520 | ||
BC050368.1 | - | 9 | 2145 | 5761074 | ||
U37791.1 | - | 9 | 2134 | |||
U38320.1 | - | 3 | 2733 | |||
U38321.1 | - | 9 | 2130 | |||
U38322.1 | - | 5 | 1941 | |||
U38431.1 | - | 9 | 2204 | |||
Y08622.1 | - | 9 | 2153 |