Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037324.2 | NP_001032401.1 | 18 | 2490 | ||
NM_001100120.1 | NP_001093590.1 | 19 | 2715 | |||
NM_001100121.1 | NP_001093591.1 | 19 | 2577 | |||
NM_014693.3 | NP_055508.3 | 19 | 2826 | |||
GenBank mRNA | AB011176.1 | - | 19 | 2572 | ||
AF192531.1 | - | 19 | 2482 | |||
AF428263.1 | - | 19 | 2754 | |||
AF428264.1 | - | 18 | 2395 | |||
AY359003.1 | - | 18 | 2446 | |||
BC142950.1 | - | 18 | 2490 | 8860334 |