Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020361.4 | NP_065094.3 | 11 | 1595 | ||
XM_011517570.2 | XP_011515872.1 | 10 | 2009 | |||
XM_017013646.1 | XP_016869135.1 | 11 | 1237 | |||
GenBank mRNA | AY044833.1 | - | 8 | 1140 | ||
BC033684.1 | - | 11 | 1555 | 5193403 | ||
BP421438.1 | - | 1 | 115 |