Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022468.4 | NP_071913.1 | 10 | 2926 | ||
XM_011522602.2 | XP_011520904.1 | 10 | 2663 | |||
XM_011522604.2 | XP_011520906.1 | 9 | 2777 | |||
XM_011522605.2 | XP_011520907.1 | 7 | 2117 | |||
GenBank mRNA | AB042328.1 | - | 10 | 2858 | ||
AJ239053.1 | - | 11 | 2410 | |||
AJ272137.1 | - | 10 | 2926 |