Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282447.2 | NP_001269376.1 | 22 | 3246 | ||
NM_025220.4 | NP_079496.1 | 22 | 3249 | |||
NM_153202.3 | NP_694882.1 | 21 | 3171 | |||
XM_005260843.1 | XP_005260900.1 | 22 | 2706 | |||
XM_006723639.1 | XP_006723702.1 | 22 | 2703 | |||
XM_006723640.1 | XP_006723703.1 | 22 | 2697 | |||
XM_011529366.1 | XP_011527668.1 | 22 | 2605 | |||
XM_011529367.1 | XP_011527669.1 | 22 | 2566 | |||
XM_011529368.1 | XP_011527670.1 | 21 | 2628 | |||
XM_011529373.1 | XP_011527675.1 | 21 | 2680 | |||
GenBank mRNA | AF466287.1 | - | 22 | 2565 | ||
AK092627.1 | - | 8 | 1443 | |||
AK094070.1 | - | 2 | 1493 | |||
AK095953.1 | - | 14 | 2351 | |||
AK123015.1 | - | 18 | 3270 | |||
AK126487.1 | - | 16 | 4247 | |||
AK126498.1 | - | 10 | 4006 | |||
AK226148.1 | - | 12 | 4310 | |||
AL117415.1 | - | 11 | 2124 | |||
AY358314.1 | - | 22 | 2676 | |||
BC125112.1 | - | 10 | 1711 | |||
BC125113.1 | - | 20 | 2247 | 40118831 |