Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001935.3 | NP_001926.2 | 26 | 3279 | ||
XM_005246371.2 | XP_005246428.1 | 26 | 2856 | |||
GenBank mRNA | BC065265.1 | - | 26 | 3279 | 6156880 | |
M74777.1 | - | 26 | 2728 | |||
X60708.1 | - | 26 | 2793 |