Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135099.1 | NP_001128571.1 | 14 | 2524 | ||
NM_005656.3 | NP_005647.3 | 14 | 2486 | |||
XM_005261043.2 | XP_005261100.1 | 13 | 2366 | |||
XM_011529733.1 | XP_011528035.1 | 12 | 2185 | |||
GenBank mRNA | AF318374.1 | - | 1 | 1579 | ||
AF329454.1 | - | 14 | 2479 | |||
AK123948.1 | - | 1 | 1820 | |||
BC051839.1 | - | 14 | 2457 | 6199625 | ||
U75329.1 | - | 13 | 2407 |