Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000540.2 | NP_000531.2 | 85 | 11857 | ||
NM_001042723.1 | NP_001036188.1 | 84 | 11842 | |||
XM_006723317.2 | XP_006723380.1 | 84 | 11789 | |||
XM_006723319.2 | XP_006723382.1 | 83 | 11774 | |||
XM_011527205.2 | XP_011525507.1 | 85 | 11807 | |||
GenBank mRNA | A20359.1 | - | 84 | 11814 | ||
AB209425.1 | - | 43 | 5099 | |||
J05200.1 | - | 84 | 11814 |