Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000726.3 | NP_000717.2 | 13 | 1231 | ||
NM_001005746.2 | NP_001005746.1 | 13 | 1181 | |||
NM_001005747.2 | NP_001005747.1 | 12 | 1319 | |||
NM_001320722.1 | NP_001307651.1 | 13 | 1479 | |||
XM_011511795.2 | XP_011510097.1 | 13 | 2948 | |||
XM_011511796.2 | XP_011510098.1 | 12 | 1902 | |||
XM_011511797.2 | XP_011510099.1 | 10 | 1951 | |||
XM_017004880.1 | XP_016860369.1 | 10 | 1043 | |||
XM_017004881.1 | XP_016860370.1 | 9 | 1111 | |||
XM_017004882.1 | XP_016860371.1 | 10 | 2436 | |||
XM_017004883.1 | XP_016860372.1 | 14 | 2770 | |||
XM_017004884.1 | XP_016860373.1 | 12 | 2604 | |||
XM_017004885.1 | XP_016860374.1 | 11 | 2444 | |||
XM_017004886.1 | XP_016860375.1 | 4 | 524 | |||
GenBank mRNA | AF038852.1 | - | 13 | 1141 | ||
AK290049.1 | - | 12 | 1300 | |||
AK291378.1 | - | 14 | 1314 | |||
AK294398.1 | - | 13 | 1155 | |||
AK316045.1 | - | 13 | 1172 | |||
AY054985.1 | - | 12 | 1290 | |||
BC075049.2 | - | 13 | 1182 | 30915578 | ||
U95020.1 | - | 13 | 1231 |