Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035.2 | NP_001026.2 | 66 | 9509 | ||
XM_006711802.3 | XP_006711865.1 | 67 | 9733 | |||
XM_006711803.3 | XP_006711866.1 | 67 | 9730 | |||
XM_006711804.3 | XP_006711867.1 | 67 | 9733 | |||
XM_006711805.3 | XP_006711868.1 | 66 | 9703 | |||
XM_006711806.3 | XP_006711869.1 | 67 | 9733 | |||
XM_006711807.3 | XP_006711870.1 | 67 | 9733 | |||
XM_006711808.3 | XP_006711871.1 | 64 | 9496 | |||
XM_006711810.3 | XP_006711873.1 | 66 | 9700 | |||
XM_017002028.1 | XP_016857517.1 | 66 | 9712 | |||
GenBank mRNA | AK294792.1 | - | 5 | 527 | ||
X98330.1 | - | 66 | 9510 |