Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001036.4 | NP_001027.3 | 8, 9 | 832 | ||
NM_001243996.2 | NP_001230925.1 | 8, 9 | 832 | |||
XM_011521880.2 | XP_011520182.1 | 8, 9 | 1020 | |||
XM_017022468.1 | XP_016877957.1 | 8, 9 | 1020 | |||
XM_017022469.1 | XP_016877958.1 | 8, 9 | 1020 | |||
XM_017022470.1 | XP_016877959.1 | 8, 9 | 3354 | |||
XM_017022471.1 | XP_016877960.1 | 8, 9 | 1020 | |||
XM_017022472.1 | XP_016877961.1 | 8, 9 | 1020 | |||
XM_017022473.1 | XP_016877962.1 | 8, 9 | 1020 | |||
XM_017022474.1 | XP_016877963.1 | 8, 9 | 984 | |||
XM_017022475.1 | XP_016877964.1 | 8, 9 | 1020 | |||
XM_017022476.1 | XP_016877965.1 | 7, 8 | 912 | |||
XM_017022477.1 | XP_016877966.1 | 8, 9 | 1021 | |||
GenBank mRNA | AB001025.1 | - | 8, 9 | 834 | ||
AJ001515.1 | - | 8, 9 | 818 |