Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002240.4 | NP_002231.1 | 4 | 2230 | ||
GenBank mRNA | D87327.1 | - | 4 | 2151 | ||
U24660.1 | - | 4 | 1860 |