Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002562.5 | NP_002553.3 | 13 | 2465 | ||
NR_033948.1 | 13 | 2688 | ||||
NR_033949.1 | 14 | 2604 | ||||
NR_033950.1 | 11 | 2565 | ||||
NR_033951.1 | 12 | 2549 | ||||
NR_033952.1 | 11 | 2476 | ||||
NR_033953.1 | 11 | 2389 | ||||
NR_033954.1 | 12 | 2368 | ||||
NR_033955.1 | 12 | 2328 | ||||
NR_033956.1 | 11 | 2255 | ||||
XM_011538419.2 | XP_011536721.1 | 12 | 2305 | |||
XM_011538420.2 | XP_011536722.1 | 11 | 1972 | |||
XM_017019364.1 | XP_016874853.1 | 13 | 2618 | |||
XM_017019365.1 | XP_016874854.1 | 12 | 2346 | |||
XM_017019366.1 | XP_016874855.1 | 12 | 2333 | |||
XM_017019367.1 | XP_016874856.1 | 11 | 2171 | |||
GenBank mRNA | BC007679.2 | - | 12 | 2267 | 3628076 |