Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OCRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000276.3 | NP_000267.2 | 11 | 1163 | ||
NM_001318784.1 | NP_001305713.1 | 11 | 1186 | |||
NM_001587.3 | NP_001578.2 | 11 | 1163 | |||
XM_005262422.2 | XP_005262479.1 | 10 | 970 | |||
XM_011531344.2 | XP_011529646.1 | 12 | 1075 | |||
XM_011531345.2 | XP_011529647.1 | 12 | 1078 | |||
XM_017029554.1 | XP_016885043.1 | 11 | 1094 | |||
GenBank mRNA | AK226116.1 | - | 11 | 1186 | ||
AK293107.1 | - | 11 | 1162 | |||
BC094726.1 | - | 11 | 1167 | 30528571 | ||
BC130612.1 | - | 11 | 1107 | 40146643 | ||
BC144106.1 | - | 11 | 1107 | 9052626 | ||
U57627.2 | - | 11 | 1163 |