Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP3CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000944.4 | NP_000935.1 | 2 | 884 | ||
NM_001130691.1 | NP_001124163.1 | 2 | 884 | |||
NM_001130692.1 | NP_001124164.1 | 2 | 884 | |||
XM_017008365.1 | XP_016863854.1 | 2 | 188 | |||
GenBank mRNA | AB451338.1 | - | 2 | 209 | ||
AB451487.1 | - | 2 | 209 | |||
AK290532.1 | - | 2 | 746 | |||
AL353950.1 | - | 2 | 615 | |||
BC025714.1 | - | 2 | 318 | 5227691 | ||
EU192652.1 | - | 2 | 884 | |||
EU192653.1 | - | 2 | 897 | |||
KU178269.1 | - | 2 | 209 | |||
KU178270.1 | - | 2 | 209 | |||
L14778.1 | - | 2 | 356 |