Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184783.2 | NP_001171712.1 | 10 | 1274 | ||
NM_001184823.1 | NP_001171752.1 | 8 | 699 | |||
NM_001324087.1 | NP_001311016.1 | 10 | 1200 | |||
NM_001324088.1 | NP_001311017.1 | 9 | 874 | |||
NM_001324089.1 | NP_001311018.1 | 10 | 978 | |||
NM_001324090.1 | NP_001311019.1 | 10 | 1142 | |||
NM_003375.4 | NP_003366.2 | 9 | 1096 | |||
GenBank mRNA | AK296635.1 | - | 9 | 790 | ||
AK315919.1 | - | 9 | 854 | |||
BC000165.2 | - | 8 | 774 | 2901130 | ||
BC012883.1 | - | 8 | 732 | 3875077 | ||
BC072407.1 | - | 8 | 764 | 6137879 | ||
BU171142.1 | - | 1 | 682 | |||
CR456964.1 | - | 6 | 582 | |||
L06328.1 | - | 7 | 677 | |||
L08666.1 | - | 9 | 907 |