Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001394.6 | NP_001385.1 | 4 | 1827 | ||
NM_057158.3 | NP_476499.1 | 5 | 2642 | |||
XM_011544428.2 | XP_011542730.1 | 4 | 4387 | |||
GenBank mRNA | AL137704.1 | - | 5 | 2642 | ||
BC002671.2 | - | 4 | 1513 | 3605895 | ||
BC014565.2 | - | 4 | 1745 | 3050199 | ||
U21108.1 | - | 4 | 1600 | |||
U48807.1 | - | 4 | 1474 |