Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001711.5 | NP_001702.1 | 8 | 1708 | ||
XM_017029724.1 | XP_016885213.1 | 7 | 1487 | |||
GenBank mRNA | AK092954.1 | - | 8 | 1487 | ||
AK093023.1 | - | 8 | 1542 | |||
AK094059.1 | - | 9 | 1517 | |||
AK222484.1 | - | 8 | 1617 | |||
AK223302.1 | - | 8 | 1588 | |||
AK297966.1 | - | 8 | 1404 | |||
AK298756.1 | - | 5 | 1039 | |||
BC002416.2 | - | 8 | 1601 | 3162633 | ||
BC004244.1 | - | 8 | 1611 | 3503374 | ||
J04599.1 | - | 8 | 1592 | |||
U11686.1 | - | 1 | 393 |