Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012128.3 | NP_036260.2 | 3 | 434 | ||
NR_024602.1 | 3 | 434 | ||||
XM_011541015.2 | XP_011539317.1 | 3 | 253 | |||
GenBank mRNA | AF127035.1 | - | 3 | 418 | ||
AK000072.1 | - | 3 | 432 | |||
AK293766.1 | - | 3 | 267 | |||
AK307785.1 | - | 3 | 378 | |||
AK314430.1 | - | 3 | 430 | |||
AY358470.1 | - | 3 | 414 | |||
BC113687.1 | - | 3 | 429 | 8322739 | ||
BC113689.1 | - | 3 | 428 | 8322741 | ||
DA668017.1 | - | 3 | 435 | |||
DA761219.1 | - | 3 | 496 |