Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMHR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164690.1 | NP_001158162.1 | 2 | 150 | ||
NM_001164691.1 | NP_001158163.1 | 2 | 150 | |||
NM_020547.2 | NP_065434.1 | 2 | 150 | |||
XM_011538173.1 | XP_011536475.1 | 2 | 150 | |||
XM_011538174.1 | XP_011536476.1 | 2 | 150 | |||
XM_011538176.1 | XP_011536478.1 | 2 | 150 | |||
XM_011538178.1 | XP_011536480.1 | 2 | 150 | |||
XM_011538179.1 | XP_011536481.1 | 2 | 150 | |||
XM_011538180.1 | XP_011536482.1 | 2 | 150 | |||
XM_011538181.1 | XP_011536483.1 | 2 | 150 | |||
XM_011538183.2 | XP_011536485.1 | 2 | 150 | |||
XM_011538184.2 | XP_011536486.1 | 2 | 150 | |||
XM_011538185.1 | XP_011536487.1 | 2 | 150 | |||
XM_011538187.2 | XP_011536489.1 | 2 | 150 | |||
XM_017019179.1 | XP_016874668.1 | 2 | 150 | |||
GenBank mRNA | AF172932.1 | - | 2 | 133 | ||
AK313593.1 | - | 2 | 139 | |||
AY714878.1 | - | 2 | 70 | |||
BC126316.1 | - | 2 | 125 | 8992032 | ||
BC136356.1 | - | 2 | 125 | 9020343 | ||
DB020573.1 | - | 2 | 150 |