Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRM SIRNA Assays ›
Gene Symbol:
PTPRM
Gene Name:
protein tyrosine phosphatase, receptor type M
Gene Aliases:
PTPRL1, R-PTP-MU, RPTPM, RPTPU, hR-PTPu
Chromosome Location:
Chr.18: 7567316 - 8406861 on Build GRCh38
UniGene ID:
Hs.49774
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
104622
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPRM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105244.1
NP_001098714.1
30
4527
NM_002845.3
NP_002836.3
28
4488
XM_011525708.2
XP_011524010.1
32
4648
XM_011525710.2
XP_011524012.1
31
4597
XM_011525712.2
XP_011524014.1
31
4573
XM_011525713.2
XP_011524015.1
32
4248
XM_011525715.2
XP_011524017.1
28
3613
XM_011525716.2
XP_011524018.1
24
3442
XM_011525717.2
XP_011524019.1
26
3322
XM_011525722.2
XP_011524024.1
26
3487
XM_017025894.1
XP_016881383.1
32
4192
XM_017025895.1
XP_016881384.1
31
4180
XM_017025896.1
XP_016881385.1
30
4153
XM_017025897.1
XP_016881386.1
31
4141
XM_017025898.1
XP_016881387.1
30
4129
XM_017025899.1
XP_016881388.1
31
4117
XM_017025900.1
XP_016881389.1
29
4102
XM_017025901.1
XP_016881390.1
29
4078
XM_017025902.1
XP_016881391.1
30
4066
XM_017025903.1
XP_016881392.1
29
4054
XM_017025904.1
XP_016881393.1
28
4027
XM_017025905.1
XP_016881394.1
31
4197
XM_017025906.1
XP_016881395.1
27
3546
XM_017025907.1
XP_016881396.1
27
3538
XM_017025908.1
XP_016881397.1
25
3523
XM_017025909.1
XP_016881398.1
25
3475
GenBank mRNA
AK294492.1
-
24
3369
BC040543.1
-
14
3761
BC051651.1
-
28
4224
5274244
BC151842.1
-
30
5073
8860327
BF725297.1
-
1
12
X58288.1
-
28
3985
Back To Top
More Information
Panther Classification:
Molecular Function -
extracellular matrix glycoprotein
protein phosphatase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of endothelial cell proliferation
protein dephosphorylation
homophilic cell adhesion via plasma membrane adhesion molecules
signal transduction
negative regulation of endothelial cell migration
retina layer formation
negative regulation of angiogenesis
neuron projection development
retinal ganglion cell axon guidance
peptidyl-tyrosine dephosphorylation
response to drug
positive regulation of vasodilation
protein tyrosine phosphatase activity
transmembrane receptor protein tyrosine phosphatase activity
protein binding
identical protein binding
cadherin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline