Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001734.3 | NP_001725.1 | 3 | 374 | ||
NM_201442.2 | NP_958850.1 | 3 | 461 | |||
XM_005253760.1 | XP_005253817.1 | 3 | 353 | |||
GenBank mRNA | AK290295.1 | - | 3 | 408 | ||
BC056903.1 | - | 3 | 401 | 6199186 | ||
CD358820.1 | - | 3 | 333 | |||
DB257339.1 | - | 3 | 374 | |||
DB265957.1 | - | 3 | 461 | |||
J04080.1 | - | 3 | 300 | |||
M18767.1 | - | 3 | 320 |