Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206838.1 | NP_001193767.1 | 30 | 5139 | ||
NM_001206839.1 | NP_001193768.1 | 29 | 5118 | |||
NM_002851.2 | NP_002842.2 | 30 | 7719 | |||
XM_005250519.1 | XP_005250576.1 | 29 | 7698 | |||
XM_017012477.1 | XP_016867966.1 | 31 | 7766 | |||
GenBank mRNA | CX872418.1 | - | 3 | 715 | ||
M93426.1 | - | 30 | 7468 |