Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNGTT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286426.1 | NP_001273355.1 | 6 | 852 | ||
NM_001286428.1 | NP_001273357.1 | 5 | 742 | |||
NM_003800.4 | NP_003791.3 | 6 | 852 | |||
XM_017011401.1 | XP_016866890.1 | 6 | 939 | |||
XM_017011402.1 | XP_016866891.1 | 6 | 939 | |||
GenBank mRNA | AB009022.1 | - | 6 | 632 | ||
AB009023.1 | - | 6 | 632 | |||
AB009024.1 | - | 6 | 632 | |||
AB012142.1 | - | 6 | 787 | |||
AB012143.1 | - | 6 | 787 | |||
AF025654.1 | - | 6 | 920 | |||
AK295719.1 | - | 7 | 730 | |||
AK299777.1 | - | 5 | 732 |