Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNGTT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286426.1 | NP_001273355.1 | 15 | 2921 | ||
NM_001286428.1 | NP_001273357.1 | 14 | 2811 | |||
NM_003800.4 | NP_003791.3 | 16 | 2990 | |||
XM_017011401.1 | XP_016866890.1 | 17 | 3095 | |||
GenBank mRNA | AF025654.1 | - | 16 | 3057 | ||
BC019954.1 | - | 16 | 2957 | 3923156 |