Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNJ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178088.1 | NP_001171559.1 | 20 | 2965 | ||
NM_003898.3 | NP_003889.1 | 21 | 3042 | |||
XM_005267198.4 | XP_005267255.1 | 20 | 2973 | |||
XM_005267199.2 | XP_005267256.1 | 20 | 2846 | |||
XM_005267200.1 | XP_005267257.1 | 10 | 1740 | |||
XM_006715592.3 | XP_006715655.1 | 21 | 3042 | |||
XM_011536224.1 | XP_011534526.1 | 21 | 3042 | |||
XM_011536225.1 | XP_011534527.1 | 21 | 3082 | |||
XM_011536226.2 | XP_011534528.1 | 15 | 3946 | |||
XM_011536227.2 | XP_011534529.1 | 21 | 3042 | |||
XM_011536228.2 | XP_011534530.1 | 15 | 4003 | |||
XM_011536230.2 | XP_011534532.1 | 21 | 3042 | |||
XM_017011406.1 | XP_016866895.1 | 20 | 3030 | |||
GenBank mRNA | AB002346.2 | - | 20 | 2778 | ||
AF039945.1 | - | 20 | 2778 | |||
AF318616.1 | - | 21 | 3019 | |||
AK074294.1 | - | 4 | 2258 | |||
AK296935.1 | - | 8 | 1147 | |||
AL157424.1 | - | 15 | 2420 | |||
AY152396.1 | - | 21 | 2944 | |||
BC043277.2 | - | 8 | 1375 | 5297160 | ||
BX648749.1 | - | 20 | 2872 |