Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018727.5 | NP_061197.4 | 16 | 3929 | ||
NM_080704.3 | NP_542435.2 | 17 | 4046 | |||
NM_080705.3 | NP_542436.2 | 16 | 3972 | |||
NM_080706.3 | NP_542437.2 | 15 | 4298 | |||
GenBank mRNA | AF196175.1 | - | 16 | 3985 | ||
AF196176.1 | - | 16 | 3970 | |||
AL136801.1 | - | 17 | 4041 |