Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CANX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024649.1 | NP_001019820.1 | 6 | 585 | ||
NM_001746.3 | NP_001737.1 | 6 | 638 | |||
XM_011534664.1 | XP_011532966.1 | 6 | 1003 | |||
XM_011534665.2 | XP_011532967.1 | 6 | 611 | |||
GenBank mRNA | AJ271880.1 | - | 5 | 477 | ||
AK129990.1 | - | 4 | 345 | |||
AK294702.1 | - | 6 | 641 | |||
AK304420.1 | - | 5 | 520 | |||
AK312334.1 | - | 6 | 545 | |||
BC003552.1 | - | 6 | 572 | 3546389 | ||
BC042843.1 | - | 6 | 519 | 4342723 | ||
L10284.1 | - | 6 | 550 | |||
L18887.1 | - | 6 | 556 | |||
M94859.1 | - | 6 | 556 | |||
S67659.1 | - | 5 | 413 |