Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004687.4 | NP_004678.3 | 19 | 4792 | ||
XM_005257784.2 | XP_005257841.1 | 18 | 4941 | |||
XM_005257785.4 | XP_005257842.1 | 20 | 4909 | |||
XM_005257786.4 | XP_005257843.1 | 19 | 5696 | |||
XM_006722168.3 | XP_006722231.1 | 19 | 4736 | |||
XM_011525460.2 | XP_011523762.1 | 20 | 5789 | |||
GenBank mRNA | AB014547.2 | - | 17 | 4682 | ||
BC035609.1 | - | 19 | 4753 | 5201989 |