Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAST SIRNA Assays ›
Gene Symbol:
CAST
Gene Name:
calpastatin
Gene Aliases:
BS-17, PLACK
Chromosome Location:
Chr.5: 96662037 - 96774683 on Build GRCh38
UniGene ID:
Hs.436186
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
10476
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAST
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042440.3
NP_001035905.1
24, 25
2188
NM_001190442.1
NP_001177371.1
25, 26
2060
NM_001284212.2
NP_001271141.1
22, 23
1836
NM_001284213.2
NP_001271142.1
21, 22
1746
NM_173060.3
NP_775083.1
23, 24
1896
NR_104285.1
11, 12
1049
XM_006714696.3
XP_006714759.1
27, 28
2202
XM_006714697.3
XP_006714760.1
26, 27
2216
XM_006714698.3
XP_006714761.1
25, 26
2180
XM_006714699.3
XP_006714762.1
26, 27
2143
XM_006714700.3
XP_006714763.1
25, 26
2162
XM_006714701.3
XP_006714764.1
25, 26
2178
XM_006714702.3
XP_006714765.1
25, 26
2054
XM_006714703.3
XP_006714766.1
24, 25
2139
XM_006714704.3
XP_006714767.1
25, 26
2087
XM_006714705.3
XP_006714768.1
24, 25
2093
XM_006714706.3
XP_006714769.1
24, 25
2014
XM_006714707.3
XP_006714770.1
24, 25
1965
XM_006714708.3
XP_006714771.1
23, 24
1925
XM_006714709.3
XP_006714772.1
23, 24
1888
XM_006714710.3
XP_006714773.1
22, 23
1843
XM_006714711.3
XP_006714774.1
22, 23
1888
XM_006714712.3
XP_006714775.1
21, 22
1835
XM_006714713.3
XP_006714776.1
11, 12
1052
XM_006714714.3
XP_006714777.1
11, 12
1049
XM_006714715.3
XP_006714778.1
12, 13
1778
XM_011543654.2
XP_011541956.1
25, 26
2138
XM_011543655.2
XP_011541957.1
25, 26
2135
XM_011543656.2
XP_011541958.1
24, 25
2082
XM_011543657.2
XP_011541959.1
24, 25
1964
XM_011543658.2
XP_011541960.1
22, 23
1817
XM_017009911.1
XP_016865400.1
25, 26
2177
XM_017009912.1
XP_016865401.1
26, 27
2188
XM_017009913.1
XP_016865402.1
26, 27
2160
XM_017009914.1
XP_016865403.1
25, 26
2159
XM_017009915.1
XP_016865404.1
26, 27
2164
XM_017009916.1
XP_016865405.1
25, 26
2175
XM_017009917.1
XP_016865406.1
25, 26
2096
XM_017009918.1
XP_016865407.1
24, 25
2123
XM_017009919.1
XP_016865408.1
24, 25
2096
XM_017009920.1
XP_016865409.1
24, 25
2006
XM_017009921.1
XP_016865410.1
24, 25
2020
XM_017009922.1
XP_016865411.1
23, 24
2057
XM_017009923.1
XP_016865412.1
23, 24
2054
XM_017009924.1
XP_016865413.1
23, 24
2036
XM_017009925.1
XP_016865414.1
23, 24
1975
XM_017009926.1
XP_016865415.1
23, 24
1972
XM_017009927.1
XP_016865416.1
24, 25
1961
XM_017009928.1
XP_016865417.1
23, 24
1960
XM_017009929.1
XP_016865418.1
23, 24
1879
XM_017009930.1
XP_016865419.1
22, 23
1849
XM_017009931.1
XP_016865420.1
22, 23
1840
XM_017009932.1
XP_016865421.1
21, 22
1903
GenBank mRNA
AB208816.1
-
25, 26
2024
AF327443.1
-
20, 21
1738
AK292289.1
-
11, 12
1052
AK292356.1
-
11, 12
1025
AK296919.1
-
25, 26
2018
AK297879.1
-
23, 24
1838
AK298541.1
-
24, 25
1965
AK299384.1
-
22, 23
1596
AK299402.1
-
24, 25
1945
AK300624.1
-
21, 22
1729
AK302206.1
-
12, 13
1052
AK314290.1
-
22, 23
1841
AK315146.1
-
22, 23
1840
AK315673.1
-
11, 12
997
AL045308.1
-
4, 5
328
AL832174.1
-
23, 24
1881
AL832349.1
-
23, 24
1884
AY189741.1
-
13, 14
1051
BC013579.1
-
24, 25
1891
3878564
BC078149.1
-
19, 20
1450
BT009783.1
-
21, 22
1673
BX648397.1
-
22, 23
1787
D16217.1
-
24, 25
1958
D50827.1
-
2, 3
206
S73329.1
-
3, 4
230
U26724.2
-
15, 16
1321
U31345.1
-
13, 14
1065
U31346.1
-
3, 4
281
U38525.1
-
6, 7
524
U58996.2
-
11, 12
1028
Back To Top
More Information
Panther Classification:
Molecular Function -
protein-binding activity modulator
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
liver development
egg activation
brain development
myoblast fusion
aging
negative regulation of endopeptidase activity
organ regeneration
myoblast differentiation
negative regulation of cell cycle arrest
cell-cell adhesion
negative regulation of type B pancreatic cell apoptotic process
protease binding
endopeptidase inhibitor activity
protein binding
calcium-dependent cysteine-type endopeptidase inhibitor activity
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-xzn5f:80/10.239.160.173:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline